Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl acetate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
830-03-5
C8H7NO4
181.15
MFCD00007326
QAUUDNIGJSLPSX-UHFFFAOYSA-N
p-nitrophenyl acetate,p-acetoxynitrobenzene,acetic acid, 4-nitrophenyl ester,p-nitrophenol acetate,acetic acid p-nitrophenyl ester,acetic acid, p-nitrophenyl ester,p-nitrophenyl acetate van,4-nitrophenyl acetate,unii-i902j0qh9s,acetic acid 4-nitrophenyl ester
13243
CHEBI:82635
(4-nitrophenyl) acetate
CC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
830-03-5
C8H7NO4
181.15
MFCD00007326
QAUUDNIGJSLPSX-UHFFFAOYSA-N
p-nitrophenyl acetate,p-acetoxynitrobenzene,acetic acid, 4-nitrophenyl ester,p-nitrophenol acetate,acetic acid p-nitrophenyl ester,acetic acid, p-nitrophenyl ester,p-nitrophenyl acetate van,4-nitrophenyl acetate,unii-i902j0qh9s,acetic acid 4-nitrophenyl ester
13243
CHEBI:82635
(4-nitrophenyl) acetate
CC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
Color Green
Infrared Spectrum Authentic
Quantity 100g
Melting Point 76.0°C to 79.0°C
Packaging Plastic bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CO2C6H4NO2
Beilstein 06, 233
Fieser 02,297
Solubility Information Solubility in water: insoluble
Formula Weight 181.15
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl acetate, 97%
Show More Show Less

EINECSNumber : 212-593-5

RTECSNumber : AJ1150000

TSCA : TSCA