Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl-beta-D-glucuronide, 99+%, ACROS Organics™

Click to view available options
Quantity:
50mg
250mg
1g
Packaging:
Glass bottle
10344-94-2
C12H13NO9
315.24
MFCD00036210
QSUILVWOWLUOEU-GOVZDWNOSA-N
4-nitrophenyl-beta-d-glucuronide,4-nitrophenylglucuronide,4-nitrophenyl beta-d-glucopyranosiduronic acid,4-nitrophenyl beta-d-glucuronide,p-nitrophenyl beta-d-glucopyranosiduronic acid,4-nitrophenyl b-d-glucosiduronic acid,p-nitrophenyl-b-d-glucuronide,p-nitrophenyl beta-d-glucuronide,beta-d-glucopyranosiduronic acid, 4-nitrophenyl, labeled with carbon-14,4-nitrophenyl-beta-d-glucopyranosiduronic acid
82565
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)C(=O)O)O)O)O
10344-94-2
C12H13NO9
315.24
MFCD00036210
QSUILVWOWLUOEU-GOVZDWNOSA-N
4-nitrophenyl-beta-d-glucuronide,4-nitrophenylglucuronide,4-nitrophenyl beta-d-glucopyranosiduronic acid,4-nitrophenyl beta-d-glucuronide,p-nitrophenyl beta-d-glucopyranosiduronic acid,4-nitrophenyl b-d-glucosiduronic acid,p-nitrophenyl-b-d-glucuronide,p-nitrophenyl beta-d-glucuronide,beta-d-glucopyranosiduronic acid, 4-nitrophenyl, labeled with carbon-14,4-nitrophenyl-beta-d-glucopyranosiduronic acid
82565
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)C(=O)O)O)O)O
Infrared Spectrum Authentic
Appearance (1% in H2O) clear colorless to faintly yellow
Packaging Glass bottle
Quantity 50mg
Water 8% max. (K.F.)
Specific Rotation −107° to −113° (20°C, 589nm) (c=1, H2O)
Color Colorless to Yellow
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 0.05% max. (p-Nitrophenol) (HPLC)
Solubility Information (1% in water) Clear colorless to light yellow
Formula Weight 315.24
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material 4-Nitrophenyl-β-D-glucuronide
Show More Show Less

MOISTURE SENSITIVE

EINECSNumber : 233-753-0