Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl-α-D-glucopyranoside, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3767-28-0
C12H15NO8
301.25
MFCD00064088
IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
4-nitrophenyl-alpha-d-glucopyranoside,4-nitrophenyl alpha-d-glucopyranoside,4-nitrophenyl a-d-glucopyranoside,4-nitrophenyl alpha-glucoside,pnpalphaglu,2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol,p-nitrophenyl alpha-d-glucopyranoside,4'-nitrophenyl-alpha-d-glucopyranoside,png,p-nitrophenyl,a-d-glucopyranoside
92969
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
3767-28-0
C12H15NO8
301.25
MFCD00064088
IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
4-nitrophenyl-alpha-d-glucopyranoside,4-nitrophenyl alpha-d-glucopyranoside,4-nitrophenyl a-d-glucopyranoside,4-nitrophenyl alpha-glucoside,pnpalphaglu,2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol,p-nitrophenyl alpha-d-glucopyranoside,4'-nitrophenyl-alpha-d-glucopyranoside,png,p-nitrophenyl,a-d-glucopyranoside
92969
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
Packaging Glass bottle
Water 0.5% max. (K.F.)
Infrared Spectrum Authentic
Appearance (0.5% aq. soln.) clear, practically colorless
Specific Rotation + 218.00
Melting Point 204.0°C to 218.0°C
Specific Rotation Condition +218.00 (20.00°C c=0.5,water)
Quantity 5g
Color White to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Solubility Information Colorless to green-yellow solution
Formula Weight 301.25
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Nitrophenyl-α-D-glucopyranoside
Show More Show Less

EINECSNumber : 223-189-3