Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl octyl ether, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
49562-76-7
C14H21NO3
251.326
MFCD00043611
WTTNDGCMXADGCJ-UHFFFAOYSA-N
4-nitrophenyl octyl ether,1-nitro-4-octyloxy benzene,p-nitrophenyl octyl ether,p-octyloxynitrobenzene,p-nitrophenoxyoctane,p-nitrophenoxvoctane,4-octyloxy-1-nitrobenzene,1-nitro-4-octyloxy benzene #,benzene,1-nitro-4-octyloxy,benzene, 1-nitro-4-octyloxy
142677
1-nitro-4-octoxybenzene
CCCCCCCCOC1=CC=C(C=C1)[N+](=O)[O-]
49562-76-7
C14H21NO3
251.326
MFCD00043611
WTTNDGCMXADGCJ-UHFFFAOYSA-N
4-nitrophenyl octyl ether,1-nitro-4-octyloxy benzene,p-nitrophenyl octyl ether,p-octyloxynitrobenzene,p-nitrophenoxyoctane,p-nitrophenoxvoctane,4-octyloxy-1-nitrobenzene,1-nitro-4-octyloxy benzene #,benzene,1-nitro-4-octyloxy,benzene, 1-nitro-4-octyloxy
142677
1-nitro-4-octoxybenzene
CCCCCCCCOC1=CC=C(C=C1)[N+](=O)[O-]
Packaging Glass bottle
Quantity 25g
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
Infrared Spectrum Authentic
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula NO2C6H4O(CH2)7CH3
Formula Weight 251.32
Physical Form Liquid After Melting
Percent Purity ≥98.5%
Chemical Name or Material 4-Nitrophenyl octyl ether, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Product may darken during storage