Alfa Aesar™ 4-Nitrophenyl phenyl ether, 99%

Manufacturer: Alfa Aesar™ B2559606

Quantity 5g

 View more versions of this product

Catalog No. AAB2559606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 620-88-2
    Molecular Formula C12H9NO3
    Formula Weight 215.21
    MDL Number MFCD00007320
    Synonym p-Nitrodiphenylether; 1-Nitro-4-phenoxybenzene
    Chemical Name or Material 4-Nitrophenyl phenyl ether

    Specifications

    Formula Weight 215.21
    Synonym p-Nitrodiphenylether; 1-Nitro-4-phenoxybenzene
    Melting Point 53° to 56°C
    TSCA TSCA
    Molecular Formula C12H9NO3
    MDL Number MFCD00007320
    CAS 620-88-2
    RTECS Number KO2680000
    Flash Point >110°C (230°F)
    Boiling Point 320°C
    Quantity 5g
    Chemical Name or Material 4-Nitrophenyl phenyl ether
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit