Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
333338-18-4
C6H16NNa2O12P
371.14
KAKKHKRHCKCAGH-UHFFFAOYSA-L
pnpp,disodium 4-nitrophenylphosphate,sodium 4-nitrophenyl phosphate,disodium 4-nitrophenyl phosphate,phosphoric acid, mono 4-nitrophenyl ester, disodium salt,disodium p-nitrophenyl phosphate,phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2,p-nitrophenyl phosphate,pnpp liquid substrate system
77949
disodium;(4-nitrophenyl) phosphate
O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
333338-18-4
C6H16NNa2O12P
371.14
KAKKHKRHCKCAGH-UHFFFAOYSA-L
pnpp,disodium 4-nitrophenylphosphate,sodium 4-nitrophenyl phosphate,disodium 4-nitrophenyl phosphate,phosphoric acid, mono 4-nitrophenyl ester, disodium salt,disodium p-nitrophenyl phosphate,phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2,p-nitrophenyl phosphate,pnpp liquid substrate system
77949
disodium;(4-nitrophenyl) phosphate
O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
Appearance (5% aq. soln.) clear
Melting Point >300.0°C
Color White to Yellow
Quantity 10g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98+%
Linear Formula O2NC6H4OP(O)(ONa)2·6H2O
Beilstein 06,237
Formula Weight 371.14
Physical Form Crystalline Powder or Crystals
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl phosphate, disodium salt, hexahydrate
Show More Show Less