Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenylacetic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
50g
100g
500g
104-03-0
C8H7NO4
181.15
MFCD00007383
YBADLXQNJCMBKR-UHFFFAOYSA-N
4-nitrophenylacetic acid,2-4-nitrophenyl acetic acid,p-nitrophenylacetic acid,4-nitrophenyl acetic acid,4-nitrobenzeneacetic acid,benzeneacetic acid, 4-nitro,2-p-nitrophenyl acetic acid,4-nitrophenylacetate,4-nitrophenylaceticacid,acetic acid, p-nitrophenyl
4661
CHEBI:40443
C1=CC(=CC=C1CC(=O)O)[N+](=O)[O-]
104-03-0
C8H7NO4
181.15
MFCD00007383
YBADLXQNJCMBKR-UHFFFAOYSA-N
4-nitrophenylacetic acid,2-4-nitrophenyl acetic acid,p-nitrophenylacetic acid,4-nitrophenyl acetic acid,4-nitrobenzeneacetic acid,benzeneacetic acid, 4-nitro,2-p-nitrophenyl acetic acid,4-nitrophenylacetate,4-nitrophenylaceticacid,acetic acid, p-nitrophenyl
4661
CHEBI:40443
C1=CC(=CC=C1CC(=O)O)[N+](=O)[O-]
Melting Point 152°C to 153°C
Infrared Spectrum Authentic
Quantity 50g
Packaging Plastic bottle
Color Beige to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula O2NC6H4CH2CO2H
Beilstein 09, 455
Merck Index 15, 6709
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,chloroform and ether
Formula Weight 181.15
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Nitrophenylacetic acid, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-168-5

RTECSNumber : AJ1130010

TSCA : TSCA