Alfa Aesar™ 4-Nitrophenylglyoxal hydrate, 97%

Manufacturer: Alfa Aesar™ H3500909

Quantity 10g

 View more versions of this product

Catalog No. AAH3500909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4974-57-6
    Formula Weight 179.13 (Anhydrous)
    MDL Number MFCD07776822
    Chemical Name or Material 4-Nitrophenylglyoxal hydrate

    Specifications

    Formula Weight 179.13 (Anhydrous)
    Melting Point 96° to 98°C
    Physical Form Crystalline Powder
    Color Cream
    MDL Number MFCD07776822
    CAS 4974-57-6
    Quantity 10g
    Chemical Name or Material 4-Nitrophenylglyoxal hydrate
    Assay 97%

    Description

    Applications
    As important industrial compounds and are mainly used as solvents. A new method for arginase assay

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit