Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Nitrophenylphosphorylcholine

Click to view available options
Quantity:
100mg
500mg
1g
21064-69-7
C11H17N2O6P
304.239
MFCD00077834
NAIXASFEPQPICN-UHFFFAOYSA-N
p-nitrophenylphosphorylcholine,o-4-nitrophenylphosphoryl choline,o-4-nitrophenylphosphoryl choline,4-nitrophenylphosphorylcholine,4-nitrophenyl 2-trimethylammonio ethyl phosphate,4-nitrophenylphorylcholine,o-4-nitrophenyl-phosphoryl choline,4-nitrophenyl 2-trimethylammonio ethyl phosphate,4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate,2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner salt
4307994
(4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate
C[N+](C)(C)CCOP(=O)([O-])OC1=CC=C(C=C1)[N+](=O)[O-]
21064-69-7
C11H17N2O6P
304.239
MFCD00077834
NAIXASFEPQPICN-UHFFFAOYSA-N
p-nitrophenylphosphorylcholine,o-4-nitrophenylphosphoryl choline,o-4-nitrophenylphosphoryl choline,4-nitrophenylphosphorylcholine,4-nitrophenyl 2-trimethylammonio ethyl phosphate,4-nitrophenylphorylcholine,o-4-nitrophenyl-phosphoryl choline,4-nitrophenyl 2-trimethylammonio ethyl phosphate,4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate,2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner salt
4307994
(4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate
C[N+](C)(C)CCOP(=O)([O-])OC1=CC=C(C=C1)[N+](=O)[O-]
Applications
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.

Solubility
Soluble in water

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Sensitivity Moisture & Light Sensitive
Quantity 500mg
Melting Point 233°C to 235°C
Color Yellow
Beilstein 1895422
Solubility Information Soluble in water
Formula Weight 304.24
Physical Form Solid
Chemical Name or Material 4-Nitrophenylphosphorylcholine

storageNote1 : -30° to -10°C