Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophthalimide, 98%, Thermo Scientific Chemicals

Catalog Number p-7081872
Click to view available options
Quantity:
5 g
25 g
89-40-7
C8H4N2O4
192.13
MFCD00005884
ANYWGXDASKQYAD-UHFFFAOYSA-N
4-nitrophthalimide, 5-nitroisoindoline-1,3-dione, phthalimide, 4-nitro, 5-nitro-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-nitro, 5-nitrophthalimide, unii-26na19ui3u, 5-nitro-2,3-dihydro-1h-isoindole-1,3-dione, ccris 4685, 5-nitro-2h-benzo c azoline-1,3-dione
6969
5-nitroisoindole-1,3-dione
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)NC2=O
89-40-7
C8H4N2O4
192.13
MFCD00005884
ANYWGXDASKQYAD-UHFFFAOYSA-N
4-nitrophthalimide, 5-nitroisoindoline-1,3-dione, phthalimide, 4-nitro, 5-nitro-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-nitro, 5-nitrophthalimide, unii-26na19ui3u, 5-nitro-2,3-dihydro-1h-isoindole-1,3-dione, ccris 4685, 5-nitro-2h-benzo c azoline-1,3-dione
6969
5-nitroisoindole-1,3-dione
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)NC2=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 200°C to 204°C
Quantity 25 g
Beilstein 180224
Formula Weight 192.13
Percent Purity 98%
Chemical Name or Material 4-Nitrophthalimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-905-5
RTECSNumber TI5625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit