Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Nitroquinoline-N-oxide, 98%, ACROS Organics™
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
Specifications
Infrared Spectrum | Authentic |
Melting Point | 154°C to 156°C |
Packaging | Glass bottle |
Quantity | 1g |
Color | Brown to Ochre-Yellow |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97.5% min. (HPLC) |
Solubility Information | Solubility in water: .. Other solubilities: soluble in acetone,dmso,dmf |
Formula Weight | 190.16 |
Show More |
Safety and Handling
GHS H Statement
May cause cancer.
GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.<
GHS Signal Word: Danger
EINECSNumber : 200-281-1