Alfa Aesar™ 4-Nitrostyrene, 98%

Manufacturer: Alfa Aesar™ H3011803

Quantity 1g

 View more versions of this product

Catalog No. AAH3011803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-13-0
    Molecular Formula C8H7NO2
    Formula Weight 149.15
    MDL Number MFCD00041254
    Chemical Name or Material 4-Nitrostyrene

    Specifications

    Formula Weight 149.15
    Molecular Formula C8H7NO2
    MDL Number MFCD00041254
    CAS 100-13-0
    Health Hazard 2 GHS H Statement
    H341-H302-H312-H332-H315-H319
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Recommended Storage −30° to −10°C
    RTECS Number WL5470000
    Flash Point 109°C (228°F)
    Boiling Point 93° to 96°C (3mmHg)
    Melting Point 25° to 28°C
    Quantity 1g
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 4-Nitrostyrene
    Assay 98%

    Description

    Applications
    Can be used to introduce amino groups into polymers. The thermal degradation of styrene-4-nitrostyrene copolymers (SNS) has been studied using differential thermal analysis (DTA) and thermogravimetry (TGA) under isothermal and dynamic conditions in dynamic nitrogen. 4—Nitrostyrene gives isomeric (4-nitropheny1)-propiona1dehydes in a 96% yield.

    Solubility
    Insoluble in water.

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air, heat. Store at 4 deg C.

    GHS H Statement
    H341-H302-H312-H332-H315-H319
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    −30° to −10°C

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit