Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-O-Methyldopamine hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
50mg
500mg
Packaging:
Glass bottle
645-33-0
C9H13NO2·HCl
203.67
MFCD00012896
KAAFITWSSODFMA-UHFFFAOYSA-N
5-2-aminoethyl-2-methoxyphenol hydrochloride, 3-hydroxy-4-methoxyphenethylamine hydrochloride, 4-o-methyldopamine hydrochloride, unii-5cp13h7ti9, 4-methoxytyramine hydrochloride, 4-methoxy-3-hydroxy-phenethylamine, 4-2-aminoethyl guaiacol hydrochloride, 4-methoxy-3-hydroxyphenethylamine hydrochloride, 4-2-aminoethyl guaiacol hcl, pubchem19594
11665606
5-(2-aminoethyl)-2-methoxyphenol;hydrochloride
COC1=C(C=C(C=C1)CCN)O.Cl
645-33-0
C9H13NO2·HCl
203.67
MFCD00012896
KAAFITWSSODFMA-UHFFFAOYSA-N
5-2-aminoethyl-2-methoxyphenol hydrochloride, 3-hydroxy-4-methoxyphenethylamine hydrochloride, 4-o-methyldopamine hydrochloride, unii-5cp13h7ti9, 4-methoxytyramine hydrochloride, 4-methoxy-3-hydroxy-phenethylamine, 4-2-aminoethyl guaiacol hydrochloride, 4-methoxy-3-hydroxyphenethylamine hydrochloride, 4-2-aminoethyl guaiacol hcl, pubchem19594
11665606
5-(2-aminoethyl)-2-methoxyphenol;hydrochloride
COC1=C(C=C(C=C1)CCN)O.Cl
Quantity 500mg
Melting Point 207°C to 211°C
Color Beige
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H3(OH)CH2CH2NH2·HCl
Formula Weight 203.67
Physical Form Fluffy Powder
Percent Purity 98%
Chemical Name or Material 4-O-Methyldopamine hydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-437-3