Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Oxo-4-(2-thienyl)butanoic acid 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
4653-08-1
C8H8O3S
184.21
MFCD00022502
ULJMYWHLMLRYSO-UHFFFAOYSA-N
3-2-thenoyl propionic acid,4-oxo-4-thiophen-2-yl butanoic acid,4-oxo-4-2-thienyl butanoic acid,4-oxo-4-2-thienyl butyric acid,3-2-thenoyl propanoic acid,4-thien-2-yl-4-oxobutyric acid,4-oxo-4-thien-2-yl butyric acid,4-thien-2-yl-4-oxobutanoic acid,4-oxo-4-thien-2-yl butanoic acid,4-oxo-4-thiophen-2-yl-butyric acid
78385
CHEBI:64434
4-oxo-4-thiophen-2-ylbutanoic acid
C1=CSC(=C1)C(=O)CCC(=O)O
4653-08-1
C8H8O3S
184.21
MFCD00022502
ULJMYWHLMLRYSO-UHFFFAOYSA-N
3-2-thenoyl propionic acid,4-oxo-4-thiophen-2-yl butanoic acid,4-oxo-4-2-thienyl butanoic acid,4-oxo-4-2-thienyl butyric acid,3-2-thenoyl propanoic acid,4-thien-2-yl-4-oxobutyric acid,4-oxo-4-thien-2-yl butyric acid,4-thien-2-yl-4-oxobutanoic acid,4-oxo-4-thien-2-yl butanoic acid,4-oxo-4-thiophen-2-yl-butyric acid
78385
CHEBI:64434
4-oxo-4-thiophen-2-ylbutanoic acid
C1=CSC(=C1)C(=O)CCC(=O)O
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Melting Point 118°C to 121°C
Color Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 184.21
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Oxo-4-(2-thienyl)butanoic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning