Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Penten-1-ol, 97%, Acros Organics

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
821-09-0
C5H10O
86.134
MFCD00002975
LQAVWYMTUMSFBE-UHFFFAOYSA-N
4-penten-1-ol,4-pentenol,4-pentenyl alcohol,2-allylethyl alcohol,unii-w0558tqc6f,4-pentene-1-ol,4-pentenol-1,4-penten-1ol,4penten-1-ol,5-hydroxy-1-pentene
13181
pent-4-en-1-ol
C=CCCCO
821-09-0
C5H10O
86.134
MFCD00002975
LQAVWYMTUMSFBE-UHFFFAOYSA-N
4-penten-1-ol,4-pentenol,4-pentenyl alcohol,2-allylethyl alcohol,unii-w0558tqc6f,4-pentene-1-ol,4-pentenol-1,4-penten-1ol,4penten-1-ol,5-hydroxy-1-pentene
13181
pent-4-en-1-ol
C=CCCCO
Packaging Glass bottle
Refractive Index 1.4289 to 1.4309 (20°C, 589nm)
Quantity 50g
Color Colorless
Infrared Spectrum Authentic
Density 0.8340g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula H2C=CH(CH2)3OH
Beilstein 01, 443
Formula Weight 86.13
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 4-Penten-1-ol, 97%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a well-ventilated place. Keep container tightly closed.

Warning