Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenolsulfonic Acid, Sodium Salt Dihydrate 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
10580-19-5,825-90-1
C6H5NaO42H2O
232.18
MFCD00150724
SXONATFNYJWFNK-UHFFFAOYSA-M
sodium 4-hydroxybenzenesulfonate dihydrate,unii-5nh81n759q,phenol-4-sulfonic acid sodium salt dihydrate,4-hydroxybenzenesulfonic acid sodium salt dihydrate,benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate,4-hydroxybenzenesulfonic acid sodium salt,sodium 4-hydroxybenzene-1-sulfonate dihydrate,phenolsulphonate sodium usp,acmc-2098hs,sodium p-phenolsulfonate dihydrate
23666329
sodium;4-hydroxybenzenesulfonate;dihydrate
C1=CC(=CC=C1O)S(=O)(=O)[O-].O.O.[Na+]
10580-19-5,825-90-1
C6H5NaO42H2O
232.18
MFCD00150724
SXONATFNYJWFNK-UHFFFAOYSA-M
sodium 4-hydroxybenzenesulfonate dihydrate,unii-5nh81n759q,phenol-4-sulfonic acid sodium salt dihydrate,4-hydroxybenzenesulfonic acid sodium salt dihydrate,benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate,4-hydroxybenzenesulfonic acid sodium salt,sodium 4-hydroxybenzene-1-sulfonate dihydrate,phenolsulphonate sodium usp,acmc-2098hs,sodium p-phenolsulfonate dihydrate
23666329
sodium;4-hydroxybenzenesulfonate;dihydrate
C1=CC(=CC=C1O)S(=O)(=O)[O-].O.O.[Na+]
Color Green to Yellow
Water 13 to 17% (K.F.)
Quantity 100g
Infrared Spectrum Authentic
Melting Point >300.0°C
Packaging Plastic bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC6H4SO3Na·2H2O
Solubility Information Solubility in water: 24g/100mL.
Formula Weight 232.18
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 4-Phenolsulfonic acid, sodium salt dihydrate, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 212-550-0

TSCA : TSCA