Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Phenoxybenzaldehyde, 98%

Click to view available options
Quantity:
1g
5g
25g
67-36-7
C13H10O2
198.221
MFCD00003383
QWLHJVDRPZNVBS-UHFFFAOYSA-N
p-phenoxybenzaldehyde,4-formyldiphenyl ether,benzaldehyde, p-phenoxy,benzaldehyde, 4-phenoxy,4-phenoxy-benzaldehyde,4-phenyloxy benzaldehyde,diphenyl ether 4-carboxaldehyde,zlchem 548,4-formyldiphenylether,4-phenoxy benzaldehyde
66139
4-phenoxybenzaldehyde
C1=CC=C(C=C1)OC2=CC=C(C=C2)C=O
67-36-7
C13H10O2
198.221
MFCD00003383
QWLHJVDRPZNVBS-UHFFFAOYSA-N
p-phenoxybenzaldehyde,4-formyldiphenyl ether,benzaldehyde, p-phenoxy,benzaldehyde, 4-phenoxy,4-phenoxy-benzaldehyde,4-phenyloxy benzaldehyde,diphenyl ether 4-carboxaldehyde,zlchem 548,4-formyldiphenylether,4-phenoxy benzaldehyde
66139
4-phenoxybenzaldehyde
C1=CC=C(C=C1)OC2=CC=C(C=C2)C=O
Applications
4-Phenoxybenzaldehyde was used in the synthesis of spirodiketopiperazine derivatives, benzoxazoles.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Density 1.132
Refractive Index 1.611
Quantity 1g
Sensitivity Air Sensitive
Flash Point >110°C (230°F)
Melting Point 24°C to 25°C
Boiling Point 184°C to 186°C (14mmHg)
Beilstein 1947841
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 4-Phenoxybenzaldehyde
Show More Show Less

EINECSNumber : 200-650-7

RTECSNumber : CU7560000

TSCA : No

Recommended Storage : Ambient temperatures