Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxybenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
67-36-7
C13H10O2
198.22
MFCD00003383
QWLHJVDRPZNVBS-UHFFFAOYSA-N
p-phenoxybenzaldehyde,4-formyldiphenyl ether,benzaldehyde, p-phenoxy,benzaldehyde, 4-phenoxy,4-phenoxy-benzaldehyde,4-phenyloxy benzaldehyde,diphenyl ether 4-carboxaldehyde,zlchem 548,4-formyldiphenylether,4-phenoxy benzaldehyde
66139
4-phenoxybenzaldehyde
C1=CC=C(C=C1)OC2=CC=C(C=C2)C=O
67-36-7
C13H10O2
198.22
MFCD00003383
QWLHJVDRPZNVBS-UHFFFAOYSA-N
p-phenoxybenzaldehyde,4-formyldiphenyl ether,benzaldehyde, p-phenoxy,benzaldehyde, 4-phenoxy,4-phenoxy-benzaldehyde,4-phenyloxy benzaldehyde,diphenyl ether 4-carboxaldehyde,zlchem 548,4-formyldiphenylether,4-phenoxy benzaldehyde
66139
4-phenoxybenzaldehyde
C1=CC=C(C=C1)OC2=CC=C(C=C2)C=O
Specific Gravity 1.132
Melting Point 24°C to 25°C
Flash Point >112°C
Boiling Point 185°C (14.0mmHg)
Density 1.1320g/mL
Refractive Index 1.6100 to 1.612
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5OC6H4CHO
Formula Weight 198.22
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 4-Phenoxybenzaldehyde, 98%
Show More Show Less

EINECSNumber : 200-650-7

RTECSNumber : CU7560000