Alfa Aesar™ 4-Phenoxybenzenesulfonamide, 98%

Manufacturer: Alfa Aesar™ H27486MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27486MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 123045-62-5
    Molecular Formula C12H11NO3S
    Formula Weight 249.29
    MDL Number MFCD00623227
    Chemical Name or Material 4-Phenoxybenzenesulfonamide

    Specifications

    Formula Weight 249.29
    Molecular Formula C12H11NO3S
    MDL Number MFCD00623227
    CAS 123045-62-5
    Melting Point 130° to 132°C
    Quantity 250mg
    Chemical Name or Material 4-Phenoxybenzenesulfonamide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit