Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxybenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2215-77-2
C13H10O3
214.22
MFCD00002539
RYAQFHLUEMJOMF-UHFFFAOYSA-N
p-phenoxybenzoic acid,benzoic acid, 4-phenoxy,diphenyl ether 4-carboxylic acid,4-phenoxy-benzoic acid,4-carboxybiphenyl ether,4-carboxydiphenyl ether,benzoic acid, p-phenoxy,p-carboxydiphenyl ether,4-phenoxy benzoic acid,4-phoc6h4cooh
75182
CHEBI:72632
4-phenoxybenzoic acid
C1=CC=C(C=C1)OC2=CC=C(C=C2)C(=O)O
2215-77-2
C13H10O3
214.22
MFCD00002539
RYAQFHLUEMJOMF-UHFFFAOYSA-N
p-phenoxybenzoic acid,benzoic acid, 4-phenoxy,diphenyl ether 4-carboxylic acid,4-phenoxy-benzoic acid,4-carboxybiphenyl ether,4-carboxydiphenyl ether,benzoic acid, p-phenoxy,p-carboxydiphenyl ether,4-phenoxy benzoic acid,4-phoc6h4cooh
75182
CHEBI:72632
4-phenoxybenzoic acid
C1=CC=C(C=C1)OC2=CC=C(C=C2)C(=O)O
Packaging Glass bottle
Color White to Yellow
Infrared Spectrum Authentic
Quantity 5g
Melting Point 160°C to 164°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5OC6H4CO2H
Formula Weight 214.22
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Phenoxybenzoic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-682-5