Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Phenoxyphenol, 99%

Click to view available options
Quantity:
5g
25g
831-82-3
C12H10O2
186.21
MFCD00002331
ZSBDGXGICLIJGD-UHFFFAOYSA-N
p-phenoxyphenol,4-hydroxydiphenyl ether,phenol, 4-phenoxy,p-hydroxydiphenyl ether,phenol, p-phenoxy,hydroquinone monophenyl ether,unii-d83r742gjr,4-phenyloxy phenol,4-phenoxy phenol,4-phenoxy-phenol
13254
CHEBI:39264
4-phenoxyphenol
C1=CC=C(C=C1)OC2=CC=C(C=C2)O
831-82-3
C12H10O2
186.21
MFCD00002331
ZSBDGXGICLIJGD-UHFFFAOYSA-N
p-phenoxyphenol,4-hydroxydiphenyl ether,phenol, 4-phenoxy,p-hydroxydiphenyl ether,phenol, p-phenoxy,hydroquinone monophenyl ether,unii-d83r742gjr,4-phenyloxy phenol,4-phenoxy phenol,4-phenoxy-phenol
13254
CHEBI:39264
4-phenoxyphenol
C1=CC=C(C=C1)OC2=CC=C(C=C2)O
Applications
4-Phenoxyphenol is a part of a group of phenolic compounds that have potential apoptotic activity, which can be used in the development of new treatments for cancer therapy.

Solubility
Soluble in water (partly).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Boiling Point 177°C to 180°C (9mmHg)
Quantity 5g
Melting Point 83°C to 85°C
Flash Point 170°C (338°F)
Beilstein 2047182
Solubility Information Soluble in water (partly).
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 4-Phenoxyphenol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 212-611-1

TSCA : Yes

Recommended Storage : Ambient temperatures