Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxyphenylboronic acid, 95+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
51067-38-0
C12H11BO3
214.03
MFCD00093312
KFXUHRXGLWUOJT-UHFFFAOYSA-N
4-phenoxyphenyl boronic acid, 4-phenoxybenzeneboronic acid, 4-phenoxyphenyl boranediol, boronic acid, 4-phenoxyphenyl, p-phenoxyphenyl boronic acid, pubchem7890, acmc-209krn, phenoxyphenylboronic acid, d0d2ja
2734377
(4-phenoxyphenyl)boronic acid
OB(O)C1=CC=C(OC2=CC=CC=C2)C=C1
51067-38-0
C12H11BO3
214.03
MFCD00093312
KFXUHRXGLWUOJT-UHFFFAOYSA-N
4-phenoxyphenyl boronic acid, 4-phenoxybenzeneboronic acid, 4-phenoxyphenyl boranediol, boronic acid, 4-phenoxyphenyl, p-phenoxyphenyl boronic acid, pubchem7890, acmc-209krn, phenoxyphenylboronic acid, d0d2ja
2734377
(4-phenoxyphenyl)boronic acid
OB(O)C1=CC=C(OC2=CC=CC=C2)C=C1
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 141°C to 145°C
Quantity 25g
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 95% min. (HPLC)
Formula Weight 214.02
Physical Form Crystalline Powder
Percent Purity 95+%
Chemical Name or Material 4-Phenoxyphenylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning