Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

6109-35-9
C15H16ClN
245.75
YYCOJKPDBCMVPP-UHFFFAOYSA-N
12999296

DA release inhibitor

Inhibitor of release of dopamine induced by methamphetamine.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES C1C(C2=CC=CC=C2CN1)C3=CC=CC=C3.Cl
IUPAC Name 4-phenyl-1,2,3,4-tetrahydroisoquinoline;hydrochloride
Formula Weight 245.75
Chemical Name or Material 4-Phenyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Grade >98%

Recommended Storage : Store at Room Temperature