Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-1,2,3-thiadiazole, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
25445-77-6
C8H6N2S
162.21
MFCD00087232
AWPNFXRMNNPKDW-UHFFFAOYSA-N
4-phenyl-1,2,3-thiadiazole, 4-phenyl-1,2,3 thiadiazole, 1,2,3-thiadiazole,4-phenyl, phenyl-thiadiazole, 1,2,3-thiadiazole, 4-phenyl, phenyl-thiadi-azole, acmc-20ambh, maybridge3_006065, 4-phenyl-1,3-thiadiazole, cambridge id 5303154
270010
4-phenylthiadiazole
C1=CC=C(C=C1)C2=CSN=N2
25445-77-6
C8H6N2S
162.21
MFCD00087232
AWPNFXRMNNPKDW-UHFFFAOYSA-N
4-phenyl-1,2,3-thiadiazole, 4-phenyl-1,2,3 thiadiazole, 1,2,3-thiadiazole,4-phenyl, phenyl-thiadiazole, 1,2,3-thiadiazole, 4-phenyl, phenyl-thiadi-azole, acmc-20ambh, maybridge3_006065, 4-phenyl-1,3-thiadiazole, cambridge id 5303154
270010
4-phenylthiadiazole
C1=CC=C(C=C1)C2=CSN=N2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 76°C to 80°C
Boiling Point ∼100°C (1.5mmHg)
Beilstein 119480
Quantity 250mg
Formula Weight 162.21
Chemical Name or Material 4-Phenyl-1,2,3-thiadiazole

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures