Alfa Aesar™ 4-Phenyl-1(2H)-phthalazinone, 98%

Manufacturer: Alfa Aesar™ H3210106

Quantity 5g

 View more versions of this product

Catalog No. AAH3210106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5004-45-5
    Molecular Formula C14H10N2O
    Formula Weight 222.25
    MDL Number MFCD00098272
    Chemical Name or Material 4-Phenyl-1(2H)-phthalazinone

    Specifications

    Formula Weight 222.25
    Molecular Formula C14H10N2O
    Melting Point 239° to 241°C
    MDL Number MFCD00098272
    CAS 5004-45-5
    Quantity 5g
    Chemical Name or Material 4-Phenyl-1(2H)-phthalazinone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit