Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-1-butyne, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
16520-62-0
C10H10
130.19
QDEOKXOYHYUKMS-UHFFFAOYSA-N
4-phenyl-1-butyne,3-butynylbenzene,but-3-yn-1-ylbenzene,benzene, 3-butynyl,2-phenylethylacetylene,ccris 7100,phenethylacetylene,4-phenyl-1-butyn,3-butynylbenzene #,4-phenylbut-1-yne
123360
but-3-ynylbenzene
C#CCCC1=CC=CC=C1
16520-62-0
C10H10
130.19
QDEOKXOYHYUKMS-UHFFFAOYSA-N
4-phenyl-1-butyne,3-butynylbenzene,but-3-yn-1-ylbenzene,benzene, 3-butynyl,2-phenylethylacetylene,ccris 7100,phenethylacetylene,4-phenyl-1-butyn,3-butynylbenzene #,4-phenylbut-1-yne
123360
but-3-ynylbenzene
C#CCCC1=CC=CC=C1
Density 0.9260g/mL
Flash Point 66°C
Boiling Point 190°C
Specific Gravity 0.926
Quantity 25g
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1-butyne

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning