Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Phenyl-2-butanone, 98%

Click to view available options
Quantity:
250g
1000g
2550-26-7
C10H12O
148.205
MFCD00008790
AKGGYBADQZYZPD-UHFFFAOYSA-N
benzylacetone,4-phenyl-2-butanone,2-butanone, 4-phenyl,4-penylbutan-2-one,1-phenyl-3-butanone,methyl phenethyl ketone,benzyl acetone,methyl 2-phenylethyl ketone,phenethyl methyl ketone,methyl phenylethyl ketone
17355
4-phenylbutan-2-one
CC(=O)CCC1=CC=CC=C1
2550-26-7
C10H12O
148.205
MFCD00008790
AKGGYBADQZYZPD-UHFFFAOYSA-N
benzylacetone,4-phenyl-2-butanone,2-butanone, 4-phenyl,4-penylbutan-2-one,1-phenyl-3-butanone,methyl phenethyl ketone,benzyl acetone,methyl 2-phenylethyl ketone,phenethyl methyl ketone,methyl phenylethyl ketone
17355
4-phenylbutan-2-one
CC(=O)CCC1=CC=CC=C1
Applications
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Boiling Point 235°C to 237°C
Quantity 250g
Flash Point 105°C (221°F)
Beilstein 1907123
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 4-Phenyl-2-butanone

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : EL9600000

TSCA : TSCA