Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Phenyl-2-thiazoleacetonitrile, 97%

Click to view available options
Quantity:
250mg
1g
41381-89-9
C11H8N2S
200.259
MFCD00551791
SOFCSIBVJROWGI-UHFFFAOYSA-N
2-4-phenylthiazol-2-yl acetonitrile,2-4-phenyl-1,3-thiazol-2-yl acetonitrile,4-phenyl-1,3-thiazol-2-yl acetonitrile,2-cyanomethyl-4-phenylthiazole,4-phenyl-thiazol-2-yl-acetonitrile,2-thiazoleacetonitrile, 4-phenyl,2-4-phenyl-1,3-thiazol-2-yl ethanenitrile,maybridge1_006883,cambridge id 5156617,4-phenyl-2-cyanomethylthiazole
704539
2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile
C1=CC=C(C=C1)C2=CSC(=N2)CC#N
41381-89-9
C11H8N2S
200.259
MFCD00551791
SOFCSIBVJROWGI-UHFFFAOYSA-N
2-4-phenylthiazol-2-yl acetonitrile,2-4-phenyl-1,3-thiazol-2-yl acetonitrile,4-phenyl-1,3-thiazol-2-yl acetonitrile,2-cyanomethyl-4-phenylthiazole,4-phenyl-thiazol-2-yl-acetonitrile,2-thiazoleacetonitrile, 4-phenyl,2-4-phenyl-1,3-thiazol-2-yl ethanenitrile,maybridge1_006883,cambridge id 5156617,4-phenyl-2-cyanomethylthiazole
704539
2-(4-phenyl-1,3-thiazol-2-yl)acetonitrile
C1=CC=C(C=C1)C2=CSC(=N2)CC#N
Quantity 250mg
Color Brown
UN Number UN3439
Formula Weight 200.26
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Phenyl-2-thiazoleacetonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.