Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-3-buten-2-one, 98+%, Acros Organics

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
122-57-6
C10H10O
146.19
MFCD00008779
BWHOZHOGCMHOBV-BQYQJAHWSA-N
benzalacetone,benzylideneacetone,methyl styryl ketone,4-phenylbut-3-en-2-one,acetocinnamone,4-phenyl-3-buten-2-one,e-4-phenylbut-3-en-2-one,benzylidene acetone,trans-benzalacetone,benzalaceton
637759
CHEBI:78399
(E)-4-phenylbut-3-en-2-one
CC(=O)C=CC1=CC=CC=C1
122-57-6
C10H10O
146.19
MFCD00008779
BWHOZHOGCMHOBV-BQYQJAHWSA-N
benzalacetone,benzylideneacetone,methyl styryl ketone,4-phenylbut-3-en-2-one,acetocinnamone,4-phenyl-3-buten-2-one,e-4-phenylbut-3-en-2-one,benzylidene acetone,trans-benzalacetone,benzalaceton
637759
CHEBI:78399
(E)-4-phenylbut-3-en-2-one
CC(=O)C=CC1=CC=CC=C1
Melting Point 38.5°C
Flash Point 116°C
Infrared Spectrum Authentic
Boiling Point 261°C
Packaging Glass bottle
Quantity 250g
Color Yellow
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula C6H5CH=CHCOCH3
Merck Index 15, 1140
Solubility Information Solubility in water: 1.3g/L (20°C). Other solubilities: sparingly soluble in petroleum ether,freely soluble in alcohol,benzene,chloroform,,ether
Formula Weight 146.19
Physical Form Low Melting Solid
Percent Purity 98+%
Chemical Name or Material 4-Phenyl-3-buten-2-one
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 204-555-1