Alfa Aesar™ 4-Phenyl-3-butyn-2-ol, 97%

Manufacturer: Alfa Aesar™ H5345014

Quantity 25g

 View more versions of this product

Catalog No. AAH5345014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5876-76-6
    Molecular Formula C10H10O
    Formula Weight 146.19
    MDL Number MFCD00046651
    Synonym 3-Hydroxy-1-phenyl-1-butyne
    Chemical Name or Material 4-Phenyl-3-butyn-2-ol

    Specifications

    Formula Weight 146.19
    Molecular Formula C10H10O
    MDL Number MFCD00046651
    CAS 5876-76-6
    Synonym 3-Hydroxy-1-phenyl-1-butyne
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point >110°C (230°F)
    Quantity 25g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.
    Chemical Name or Material 4-Phenyl-3-butyn-2-ol
    Assay 97%

    GHS H Statement
    H301-H319
    Toxic if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P321-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit