Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-4-piperidinecarboxylic Acid p-methylbenzenesulfonate 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
83949-32-0
C12H15NO2·C7H8O3S
377.45
MFCD00044709
NQLZTDKDXBKUGY-UHFFFAOYSA-N
4-methylbenzene-1-sulfonic acid; 4-phenylpiperidine-4-carboxylic acid,4-carboxy-4-phenylpiperidine tosylate,4-phenylpiperidine-4-carboxylic acid compound with 4-methylbenzenesulfonic acid 1:1,4-phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate,4-phenyl-4-piperidinecarboxylic acid 4-methylbenzenesulfonate,4-phenylpiperidine-4-carboxylic acid 4-toluenesulfonate,isonipecotic acid, 4-phenyl-; para-toluene sulfonate,4-phenyl-4-carboxy piperidine tosylate,4-carboxy-4-phenylpiperidine p-toluenesulfonate,4-phenyl-piperidine-4-carboxylic acid tosylate
2723657
4-methylbenzenesulfonic acid;4-phenylpiperidine-4-carboxylic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1CNCCC1(C2=CC=CC=C2)C(=O)O
83949-32-0
C12H15NO2·C7H8O3S
377.45
MFCD00044709
NQLZTDKDXBKUGY-UHFFFAOYSA-N
4-methylbenzene-1-sulfonic acid; 4-phenylpiperidine-4-carboxylic acid,4-carboxy-4-phenylpiperidine tosylate,4-phenylpiperidine-4-carboxylic acid compound with 4-methylbenzenesulfonic acid 1:1,4-phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate,4-phenyl-4-piperidinecarboxylic acid 4-methylbenzenesulfonate,4-phenylpiperidine-4-carboxylic acid 4-toluenesulfonate,isonipecotic acid, 4-phenyl-; para-toluene sulfonate,4-phenyl-4-carboxy piperidine tosylate,4-carboxy-4-phenylpiperidine p-toluenesulfonate,4-phenyl-piperidine-4-carboxylic acid tosylate
2723657
4-methylbenzenesulfonic acid;4-phenylpiperidine-4-carboxylic acid
CC1=CC=C(C=C1)S(=O)(=O)O.C1CNCCC1(C2=CC=CC=C2)C(=O)O
Melting Point 225°C to 229°C
Packaging Glass bottle
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: 65g/L methanol- 14g/L ethanol,3.3g/L 2-propanol- 4.4g/L acetone,1.9g/L methylene chloride,100g/L n,n-dimethylformamide
Formula Weight 377.45
Chemical Name or Material 4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 281-391-7