Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Phenylamino)biphenyl, 98%

Click to view available options
Quantity:
250mg
1g
5g
32228-99-2
C18H15N
245.325
MFCD01318677
YGNUPJXMDOFFDO-UHFFFAOYSA-N
n-phenyl-4-biphenylamine,n-phenyl-1,1'-biphenyl-4-amine,1,1'-biphenyl-4-amine, n-phenyl,n-phenylbiphenyl-4-amine,ethyl propriolate,4-anilinobiphenyl,biphenyl-4-yl-phenylamine,1,1'-biphenyl-4-amine,n-phenyl,acmc-209hrz,biphenyl-4-yl-phenylamine
13087229
N,4-diphenylaniline
C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=CC=C3
32228-99-2
C18H15N
245.325
MFCD01318677
YGNUPJXMDOFFDO-UHFFFAOYSA-N
n-phenyl-4-biphenylamine,n-phenyl-1,1'-biphenyl-4-amine,1,1'-biphenyl-4-amine, n-phenyl,n-phenylbiphenyl-4-amine,ethyl propriolate,4-anilinobiphenyl,biphenyl-4-yl-phenylamine,1,1'-biphenyl-4-amine,n-phenyl,acmc-209hrz,biphenyl-4-yl-phenylamine
13087229
N,4-diphenylaniline
C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=CC=C3
Sensitivity Air Sensitive
Quantity 5g
Beilstein 2806457
Formula Weight 245.32
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-(Phenylamino)biphenyl