Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-(Phenylazo)diphenylamine, 97%, Acros Organics™
Click to view available options
Quantity:
100g
Specifications
Infrared Spectrum | Authentic |
Melting Point | 84.0°C to 91.0°C |
Quantity | 100g |
Assay Percent Range | 96% min. (HPLC) |
Linear Formula | C6H5N=NC6H4NHC6H5 |
Beilstein | 16,314 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether |
Formula Weight | 273.33 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Phenylazo)diphenylamine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Warning
EINECSNumber : 202-972-3