Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Phenylazophenol, 98%

Click to view available options
Quantity:
5g
25g
100g
1689-82-3
C12H10N2O
198.225
MFCD00002330
JTSBGMZPPPULTA-UHFFFAOYSA-N
4-hydroxyazobenzene,4-phenylazophenol,4-phenyldiazenyl phenol,solvent yellow 7,p-phenylazophenol,4-phenylazo phenol,p-hydroxyazobenzene,organol yellow ap,c.i. solvent yellow 7,phenol, 4-phenylazo
5354198
4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
C1=CC=C(C=C1)NN=C2C=CC(=O)C=C2
1689-82-3
C12H10N2O
198.225
MFCD00002330
JTSBGMZPPPULTA-UHFFFAOYSA-N
4-hydroxyazobenzene,4-phenylazophenol,4-phenyldiazenyl phenol,solvent yellow 7,p-phenylazophenol,4-phenylazo phenol,p-hydroxyazobenzene,organol yellow ap,c.i. solvent yellow 7,phenol, 4-phenylazo
5354198
4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
C1=CC=C(C=C1)NN=C2C=CC(=O)C=C2
Applications
4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.

Solubility
Soluble in acetone, ethanol, benzene and ether. Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Quantity 5g
Melting Point 152°C to 155°C
Formula Weight 198.23
Percent Purity 98%
Chemical Name or Material 4-Phenylazophenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SM8300000

TSCA : TSCA