Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylbutyric acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1821-12-1
C10H12O2
164.2
OBKXEAXTFZPCHS-UHFFFAOYSA-N
4-phenylbutyric acid,benzenebutanoic acid,benzenebutyric acid,phenylbutyrate,gamma-phenylbutyric acid,omega-phenylbutanoic acid,phenylbutyric acid,4-phenylbutyrate,butyric acid, 4-phenyl,4-phenyl-n-butyric acid
4775
CHEBI:41500
4-phenylbutanoic acid
C1=CC=C(C=C1)CCCC(=O)O
1821-12-1
C10H12O2
164.2
OBKXEAXTFZPCHS-UHFFFAOYSA-N
4-phenylbutyric acid,benzenebutanoic acid,benzenebutyric acid,phenylbutyrate,gamma-phenylbutyric acid,omega-phenylbutanoic acid,phenylbutyric acid,4-phenylbutyrate,butyric acid, 4-phenyl,4-phenyl-n-butyric acid
4775
CHEBI:41500
4-phenylbutanoic acid
C1=CC=C(C=C1)CCCC(=O)O
Quantity 25g
Color White to Yellow
Melting Point 49.0°C to 52.0°C
Packaging Glass bottle
Flash Point >110°C
Boiling Point 165.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H5(CH2)3CO2H
Beilstein 09,539
Solubility Information Solubility in water: 5.3g/L water (40 degr. c)
Formula Weight 164.2
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Phenylbutyric acid
Show More Show Less