Alfa Aesar™ 4-Phenylbutyryl chloride, 97%

Manufacturer: Alfa Aesar™ H5038806

Quantity 5g

 View more versions of this product

Catalog No. AAH5038806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18496-54-3
    Molecular Formula C10H11ClO
    Linear Formula C6H5CH2CH2CH2COCl
    Formula Weight 182.65
    MDL Number MFCD00798080
    UN Number UN3265
    Chemical Name or Material 4-Phenylbutyryl chloride

    Specifications

    Formula Weight 182.65
    Molecular Formula C10H11ClO
    Linear Formula C6H5CH2CH2CH2COCl
    MDL Number MFCD00798080
    CAS 18496-54-3
    UN Number UN3265
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Boiling Point 273°C
    Quantity 5g
    Chemical Name or Material 4-Phenylbutyryl chloride
    Assay 97%

    Description

    Applications
    4-Phenylbutyryl chloride is used as organic chemical synthesis intermediate.

    Solubility
    It reacts with water.

    Notes
    Moisture sensitive. Incompatible with oxidizing agents, bases, active metals, alcohols and amines.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit