Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylcyclohexanone, 98+%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
4894-75-1
C12H14O
174.24
MFCD00001641
YKAYMASDSHFOGI-UHFFFAOYSA-N
4-phenylcyclohexanone,cyclohexanone, 4-phenyl,4-phenyl-1-cyclohexanone,4-phenyl cyclohexanone,4-phenyl-cyclohexanone,4-phenylcyclo-hexanone,acmc-1aoge,enamine_005496,1-n-phenylpiperidin-4-one,4-phenyl-cyclohexan-1-one
78605
4-phenylcyclohexan-1-one
C1CC(=O)CCC1C2=CC=CC=C2
4894-75-1
C12H14O
174.24
MFCD00001641
YKAYMASDSHFOGI-UHFFFAOYSA-N
4-phenylcyclohexanone,cyclohexanone, 4-phenyl,4-phenyl-1-cyclohexanone,4-phenyl cyclohexanone,4-phenyl-cyclohexanone,4-phenylcyclo-hexanone,acmc-1aoge,enamine_005496,1-n-phenylpiperidin-4-one,4-phenyl-cyclohexan-1-one
78605
4-phenylcyclohexan-1-one
C1CC(=O)CCC1C2=CC=CC=C2
Flash Point >100°C
Color White
Packaging Glass bottle
Melting Point 77°C to 80°C
Quantity 10g
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula C6H5C6H9(=O)
Beilstein 07, III, 1466
Solubility Information Solubility in water: slightly soluble
Formula Weight 174.24
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 4-Phenylcyclohexanone
Show More Show Less

EINECSNumber : 225-517-