Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylcyclohexanone 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
4894-75-1
C12H14O
174.243
MFCD00001641
YKAYMASDSHFOGI-UHFFFAOYSA-N
4-phenylcyclohexanone, cyclohexanone, 4-phenyl, 4-phenyl-1-cyclohexanone, 4-phenyl cyclohexanone, 4-phenyl-cyclohexanone, 4-phenylcyclo-hexanone, acmc-1aoge, enamine_005496, 1-n-phenylpiperidin-4-one, 4-phenyl-cyclohexan-1-one
78605
4-phenylcyclohexan-1-one
C1CC(=O)CCC1C2=CC=CC=C2
4894-75-1
C12H14O
174.243
MFCD00001641
YKAYMASDSHFOGI-UHFFFAOYSA-N
4-phenylcyclohexanone, cyclohexanone, 4-phenyl, 4-phenyl-1-cyclohexanone, 4-phenyl cyclohexanone, 4-phenyl-cyclohexanone, 4-phenylcyclo-hexanone, acmc-1aoge, enamine_005496, 1-n-phenylpiperidin-4-one, 4-phenyl-cyclohexan-1-one
78605
4-phenylcyclohexan-1-one
C1CC(=O)CCC1C2=CC=CC=C2
Color White
Melting Point 78°C
Quantity 250g
Formula Weight 174.24
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Phenylcyclohexanone