Alfa Aesar™ 4-(Phenylethynyl)benzeneboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H5169906

Quantity 5g

 View more versions of this product

Catalog No. AAH5169906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1190376-20-5
    Molecular Formula C20H21BO2
    Formula Weight 304.19
    MDL Number MFCD16294498
    Beilstein 20037873
    Synonym Diphenylacetylene-4-boronic acid pinacol ester; 4,4,5,5-Tetramethyl-2-(4-phenylethynyl)-1,3,2-dioxaborolane
    Chemical Name or Material 4-(Phenylethynyl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 304.19
    Molecular Formula C20H21BO2
    MDL Number MFCD16294498
    CAS 1190376-20-5
    Synonym Diphenylacetylene-4-boronic acid pinacol ester; 4,4,5,5-Tetramethyl-2-(4-phenylethynyl)-1,3,2-dioxaborolane
    Quantity 5g
    Beilstein 20037873
    Chemical Name or Material 4-(Phenylethynyl)benzeneboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit