Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Phenylpiperidine, 96%

Click to view available options
Quantity:
1g
5g
771-99-3
C11H15N
161.248
MFCD00006002
UTBULQCHEUWJNV-UHFFFAOYSA-N
4-phenyl-piperidine,piperidine, 4-phenyl,4-phenyl piperidine,4-phenyl piperidine,chembl20969,4-phenypiperdine,4-phenyl piperidin,4-phenyl piperidine,pubchem8009,4-phenylpiperidine
69873
4-phenylpiperidine
C1CNCCC1C2=CC=CC=C2
771-99-3
C11H15N
161.248
MFCD00006002
UTBULQCHEUWJNV-UHFFFAOYSA-N
4-phenyl-piperidine,piperidine, 4-phenyl,4-phenyl piperidine,4-phenyl piperidine,chembl20969,4-phenypiperdine,4-phenyl piperidin,4-phenyl piperidine,pubchem8009,4-phenylpiperidine
69873
4-phenylpiperidine
C1CNCCC1C2=CC=CC=C2
Applications
4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Quantity 1g
Color Yellow to Brown
Melting Point 61°C to 64°C
Beilstein 124508
Formula Weight 161.24
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 4-Phenylpiperidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning