Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylpiperidine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
771-99-3
C11H15N
161.25
MFCD00006002
UTBULQCHEUWJNV-UHFFFAOYSA-N
4-phenyl-piperidine,piperidine, 4-phenyl,4-phenyl piperidine,chembl20969,4-phenypiperdine,4-phenyl piperidin,pubchem8009,4-phenylpiperidine
69873
4-phenylpiperidine
C1CNCCC1C2=CC=CC=C2
771-99-3
C11H15N
161.25
MFCD00006002
UTBULQCHEUWJNV-UHFFFAOYSA-N
4-phenyl-piperidine,piperidine, 4-phenyl,4-phenyl piperidine,chembl20969,4-phenypiperdine,4-phenyl piperidin,pubchem8009,4-phenylpiperidine
69873
4-phenylpiperidine
C1CNCCC1C2=CC=CC=C2
Boiling Point 255.0°C to 257.0°C (727.0mmHg)
Color White to Brown
Quantity 5g
Packaging Glass bottle
Melting Point 57.0°C to 63.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 20, 291
Formula Weight 161.25
Physical Form Crystalline Powder and/or Crystals
Percent Purity 98%
Chemical Name or Material 4-Phenylpiperidine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-243-1