Alfa Aesar™ 4-Phenylsemicarbazide, 98+%

Manufacturer: Alfa Aesar™ B2557818

Quantity 50g

 View more versions of this product

Catalog No. AAB2557818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 537-47-3
    Molecular Formula C6H5NHCONHNH2
    Formula Weight 151.17
    MDL Number MFCD00007590
    Beilstein 511107
    Merck Index 14,7312
    Synonym N-Phenylhydrazinecarboxamide; Anilinoformylhydrazine
    Chemical Name or Material 4-Phenylsemicarbazide

    Specifications

    Formula Weight 151.17
    Molecular Formula C6H5NHCONHNH2
    Melting Point 123° to 125°C
    MDL Number MFCD00007590
    CAS 537-47-3
    Beilstein 511107
    Merck Index 14,7312
    Synonym N-Phenylhydrazinecarboxamide; Anilinoformylhydrazine
    Quantity 50g
    Chemical Name or Material 4-Phenylsemicarbazide
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit