Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Phenylsemicarbazide, 98+%

Click to view available options
Quantity:
10g
25g
50g
537-47-3
C7H9N3O
151.169
MFCD00007590
MOCKWYUCPREFCZ-UHFFFAOYSA-N
4-phenylsemicarbazide,n-phenylhydrazinecarboxamide,3-amino-1-phenylurea,hydrazinecarboxamide, n-phenyl,anilinoformylhydrazine,unii-bpu96sx57a,semicarbazide, 4-phenyl,bpu96sx57a,hydrazinecarboxanilide,anilinocarbonylhydrazine
10837
1-amino-3-phenylurea
C1=CC=C(C=C1)NC(=O)NN
537-47-3
C7H9N3O
151.169
MFCD00007590
MOCKWYUCPREFCZ-UHFFFAOYSA-N
4-phenylsemicarbazide,n-phenylhydrazinecarboxamide,3-amino-1-phenylurea,hydrazinecarboxamide, n-phenyl,anilinoformylhydrazine,unii-bpu96sx57a,semicarbazide, 4-phenyl,bpu96sx57a,hydrazinecarboxanilide,anilinocarbonylhydrazine
10837
1-amino-3-phenylurea
C1=CC=C(C=C1)NC(=O)NN
Melting Point 123°C to 125°C
Quantity 25g
Beilstein 511107
Merck Index 14,7312
Formula Weight 151.17
Percent Purity ≥98%
Chemical Name or Material 4-Phenylsemicarbazide