Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Picolyl chloride hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
100g
Packaging:
Glass bottle
1822-51-1
C6H6ClN·HCl
164.04
MFCD00012826
ZDHKVKPZQKYREU-UHFFFAOYSA-N
4-chloromethyl pyridine hydrochloride,4-picolyl chloride hydrochloride,4-chloromethyl pyridinium chloride,4-chloromethylpyridine hydrochloride,4-chloromethyl-pyridine hydrochloride,ccris 1728,isonicotinyl chloride hydrochloride,pyridine, 4-chloromethyl-, hydrochloride,4-chloromethyl pyridine hcl,4-chloromethyl pyridine, chloride
74570
4-(chloromethyl)pyridine;hydrochloride
C1=CN=CC=C1CCl.Cl
1822-51-1
C6H6ClN·HCl
164.04
MFCD00012826
ZDHKVKPZQKYREU-UHFFFAOYSA-N
4-chloromethyl pyridine hydrochloride,4-picolyl chloride hydrochloride,4-chloromethyl pyridinium chloride,4-chloromethylpyridine hydrochloride,4-chloromethyl-pyridine hydrochloride,ccris 1728,isonicotinyl chloride hydrochloride,pyridine, 4-chloromethyl-, hydrochloride,4-chloromethyl pyridine hcl,4-chloromethyl pyridine, chloride
74570
4-(chloromethyl)pyridine;hydrochloride
C1=CN=CC=C1CCl.Cl
Melting Point 160°C to 163°C
Packaging Glass bottle
Quantity 100g
Color Yellow to Orange
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Fieser 04,391
Solubility Information Solubility in water: 750g/L (20°C)
Formula Weight 164.04
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Picolyl chloride hydrochloride, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 217-350-7