Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Piperidinemethanol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
6457-49-4
C6H13NO
115.17
MFCD00174228
XBXHCBLBYQEYTI-UHFFFAOYSA-N
4-piperidinemethanol,4-hydroxymethyl piperidine,4-piperidylmethanol,4-hydroxymethylpiperidine,piperidin-4-yl methanol,piperidin-4-yl-methanol,piperidine-4-methanol,4-piperidylmethan-1-ol,4-piperidine methanol,4-piperidyl carbinol
420771
piperidin-4-ylmethanol
C1CNCCC1CO
6457-49-4
C6H13NO
115.17
MFCD00174228
XBXHCBLBYQEYTI-UHFFFAOYSA-N
4-piperidinemethanol,4-hydroxymethyl piperidine,4-piperidylmethanol,4-hydroxymethylpiperidine,piperidin-4-yl methanol,piperidin-4-yl-methanol,piperidine-4-methanol,4-piperidylmethan-1-ol,4-piperidine methanol,4-piperidyl carbinol
420771
piperidin-4-ylmethanol
C1CNCCC1CO
Boiling Point 118.0°C to 120.0°C (10.0 mmHg)
Color White
Quantity 100g
Flash Point >110°C
Packaging Glass bottle
Melting Point 57.0°C to 61.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 115.17
Physical Form Crystalline Powder, Chunks and/or Solid
Percent Purity 98%
Chemical Name or Material 4-Piperidinemethanol
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

Danger