Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Piperidone ethylene ketal, 98+%

Manufacturer:  Alfa Aesar™ A1241918

Quantity 50g

 View more versions of this product

Catalog No. AAA1241918


  Check Availability
Add to cart

Specifications

Description

Applications
4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

Solubility
Soluble in water (Partly miscible).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Description & Specifications

Chemical Identifiers

177-11-7
143.19
506799
4-Piperidone ethylene ketal
C7H13NO2
MFCD00005976
1,4-Dioxa-8-azaspiro[4.5]decane

Specifications

81°C (177°F)
50g
C7H13NO2
MFCD00005976
1,4-Dioxa-8-azaspiro[4.5]decane
≥98%
99° to 101°C (14mmHg)
177-11-7
143.19
506799
4-Piperidone ethylene ketal

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit