4-PPBP maleate, Tocris

Specific σ ligand, NMDA antagonist

Manufacturer: R&D Systems 0620/10

Quantity 10mg

 View more versions of this product

Catalog No. 06-201-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C21H27N·C4H4O4
    Formula Weight 409.52
    Synonym 4-Phenyl-1-(4-phenylbutyl)piperidine maleate
    Chemical Name or Material 4-PPBP maleate

    Specifications

    Quantity 10mg
    Formula Weight 409.52
    Assay Percent Range >99%
    Molecular Formula C21H27N·C4H4O4
    CAS 201216-39-9
    Synonym 4-Phenyl-1-(4-phenylbutyl)piperidine maleate
    Recommended Storage Store at Room Temperature
    Chemical Name or Material 4-PPBP maleate

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit