Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propoxyphenol, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
18979-50-5
C9H12O2
152.193
MFCD00002335
KIIIPQXXLVCCQP-UHFFFAOYSA-N
phenol, 4-propoxy,4-n-propoxyphenol,p-propoxyphenol,phenol, p-propoxy,hydroquinone monopropyl ether,p-propoxy phenol,4-propyloxyphenol,4-propoxy-phenol,pubchem13204,para-propoxyphenol
29352
4-propoxyphenol
CCCOC1=CC=C(C=C1)O
18979-50-5
C9H12O2
152.193
MFCD00002335
KIIIPQXXLVCCQP-UHFFFAOYSA-N
phenol, 4-propoxy,4-n-propoxyphenol,p-propoxyphenol,phenol, p-propoxy,hydroquinone monopropyl ether,p-propoxy phenol,4-propyloxyphenol,4-propoxy-phenol,pubchem13204,para-propoxyphenol
29352
4-propoxyphenol
CCCOC1=CC=C(C=C1)O
Flash Point >110°C
Melting Point 55.0°C to 58.0°C
Packaging Glass bottle
Quantity 25g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula CH3CH2CH2OC6H4OH
Formula Weight 152.19
Percent Purity 96%
Chemical Name or Material 4-Propoxyphenol, 96%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA