Learn More
Accela Chembio IncSupplier Diversity Partner 4-PYRAZOLEBORONIC ACID PI 5G  DFS Item
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY0042185G
4-PYRAZOLEBORONIC ACID PINACOL ESTER, 5G, 269410-08-4, MFCD03453063, 97+%, RETEST PERIOD: 540DAYS, LIGHT SENSITIVE/MOISTURE SENSITIVE/NITROGEN OR ARGON/+4 c
Catalog No. 50-648-172