Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Pyridineacetonitrile, ≥97%

Click to view available options
Quantity:
250mg
1g
13121-99-8
C7H6N2
118.139
MFCD03412601
BMVSAKPRNWZCPG-UHFFFAOYSA-N
pyridin-4-yl-acetonitrile,2-pyridin-4-yl acetonitrile,4-pyridylacetonitrile,4-pyridineacetonitrile,pyridin-4-ylacetonitrile,pyridine-4-acetonitrile,rarechem ak ml 0204,2-4-pyridyl acetonitrile,4-pyridylacetonitrilehydrochloride,4-cyanomethyl-pyridine
4112085
2-pyridin-4-ylacetonitrile
C1=CN=CC=C1CC#N
13121-99-8
C7H6N2
118.139
MFCD03412601
BMVSAKPRNWZCPG-UHFFFAOYSA-N
pyridin-4-yl-acetonitrile,2-pyridin-4-yl acetonitrile,4-pyridylacetonitrile,4-pyridineacetonitrile,pyridin-4-ylacetonitrile,pyridine-4-acetonitrile,rarechem ak ml 0204,2-4-pyridyl acetonitrile,4-pyridylacetonitrilehydrochloride,4-cyanomethyl-pyridine
4112085
2-pyridin-4-ylacetonitrile
C1=CN=CC=C1CC#N
Applications
A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
UN Number UN3439
Beilstein 471559
Formula Weight 118.14
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material 4-Pyridineacetonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.